missing translation for 'onlineSavingsMsg'
Learn More

PKCθ (pT538) Mouse anti-Human, Unlabeled, Clone: 19, BD

Mouse Monoclonal Antibody

494.34 €

Specifications

Antigen PKCθ (pT538)
Clone 19
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15816279
View Documents
BD Biosciences
612735
150 μg
ONLINE EXCLUSIVE PRICE
515.00 €
494.34 € / Each
Save 20.66 €
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Protein Kinase C (PKC) family of homologous serine/threonine protein kinases is involved in a number of processes such as growth, differentiation, and cytokine secretion. Three categories exist, conventional PKC (cPKC), novel PKC (nPKC), and atypical PKC (aPKC). These proteins are products of multiple genes and alternative splicing and have different modes of activation. For example, cPKC's members (α, βI, βII, and γ) are calcium activated, phospholipid-dependent serine/threonine specific enzymes which can also be activated by phorbol esters. However, the novel PKC (nPKC) subfamily members (δ , ε, π, and θ isoforms) and the atypical PKC (PKC) subfamily members (ζ , f , andλ isoforms) are Ca2+; independent. The aPKC members are unique in that their activity is independent of diacylglycerols and phorbol esters. The PKC pathway represents a major signal transduction system that is activated following ligand-stimulation of transmembrane receptors by hormones, neurotransmitters and growth factors. PKCθ transcripts are expressed in most tissues with the highest levels being found in hematopoietic tissues and cell lines, including T cells and thymocytes. PKCθ RNA is readily detectable in skeletal muscle, lung, and brain. However, PKCθ expression is not detected in several human carcinoma cell lines. Abundant expression of this PKC isozyme in hematopoietic cells suggests that it may have a role in growth and differentiation processes of these cells. The 9/PKC monoclonal antibody recognizes the phosphorylated threonine 538 (pT538) of human PKCθ.

Western Blotting

Specifications

Specifications

PKCθ (pT538)
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG2a
Affinity Purified
19
Western Blot
Unconjugated
RUO
Human
Phosphorylated Human PKCθ Peptide
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.