missing translation for 'onlineSavingsMsg'
Learn More

Rabaptin-5 Mouse, Unlabeled, Clone: 20, BD

Product Code. 15855579
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15855579 50 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15855579 Supplier BD Biosciences Supplier No. 610676

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Rabs are small GTP-binding proteins that are localized to specific intracellular vesicles and organelles. They are important for the regulation of vesicular traffic. It is thought that Rabs cycle between GTP- and GDP-bound forms and this cycle is related to their function. Rabaptin-5 is protein that interacts with GTP-Rab5. Although Rabaptin-5 is localized in the cytosol, it is also recruited to the endosomal fraction in a Rab5 and GTP-dependent manner. The removal of Rabaptin-5 from the cytosol substantially impairs GTP and Rab5-dependent endosomal fusion. This indicates that Rabaptin-5 may be an important partner of Rab5 during endocytosis. Rabaptin-5 contains 862 amino acids and both ends of the protein contain coiled-coil domains typically found in vesicular transport proteins.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabaptin-5
Applications Western Blot
Classification Monoclonal
Clone 20
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.