missing translation for 'onlineSavingsMsg'
Learn More

Rac1 Mouse, Unlabeled, Clone: 102, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610651

 View more versions of this product

Product Code. 15815569

  • ONLINE EXCLUSIVE PRICE
    371.00 €
    355.24 € / 1 set
    Save 15.76 €
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Rac1 is a member of the expanding family of Ras-related GTPase proteins. These small proteins have the ability to switch between two different conformational states: the active (GTP-bound) and the inactive (GDP-bound). The GTPase activity is increased by specific activator proteins named GAPs. Rac1, like many of its relatives, contains the consensus sequence Cys-X-X-X-COOH, which localizes it to the plasma membrane. Growth factor stimulation of cells results in a dramatic reorganization of the actin filament network and membrane ruffling, processes for which Rac1 is essential. Constitutive expression of Rac1 leads to cell transformation. This process is mediated by the Rac1-dependent activation of the p65PAK serine/threonine kinase. In addition, IQGAP1 has been identified as a GTPase activating protein specific for Rac1. Due to sequence homology among various small GTPase family members, potential cross-reactivity could be observed with this antibody. BD Biosciences Pharmingen, however, has not performed such cross-reactivity testing.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rac1
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
150 μg
Cell Biology
Canine, Chicken, Human, Mouse, Rat
IgG2b κ
Western Blot
102
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.