Learn More
RASEF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531016
Code : A0
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A431. Predicted reactivity: Mouse (85%), Rat (84%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RASEF Polyclonal antibody specifically detects RASEF in Human samples. It is validated for Western Blot.Specifications
RASEF | |
Polyclonal | |
Unconjugated | |
RASEF | |
BC030882; RAB45; RAS and EF hand domain containing; RAS and EF-hand domain containing; ras and EF-hand domain-containing protein; Ras and EF-hand domain-containing protein homolog; RASEF; ras-related protein Rab-45 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
158158 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8IZ41 | |
RASEF | |
Recombinant fragment corresponding to a region within amino acids 507 and 740 of Human RASEF | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |