Learn More
RBP4, Human, Matched Antibody Pair (AP44), Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00005950-AP44.1Set
Product Code. 16102554
  • 835.00 € / 1 set
Includes: Capture antibody: mouse monoclonal anti-RBP4, IgG1 Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-RBP4, IgG1 Lambda (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
RBP4
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human RBP4.
RBP4
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-RBP4, IgG1 Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-RBP4, IgG1 Lambda (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
ELISA
Biotin
RBP4
Mouse
RUO
5950