missing translation for 'onlineSavingsMsg'
Learn More

RIP Mouse, Unlabeled, Clone: 38, BD

Product Code. 15815479
Change view
Click to view available options
Quantity:
150 μg
50 μg
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15815479 150 μg 1 set
15805479 50 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15815479 Supplier BD Biosciences Supplier No. 610459

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Binding or cross linking of the Fas antigen (also known as APO-1 and CD95) is known to elicit apoptosis in susceptible cells. Fas is a member of a family of cell surface receptors which includes tumor necrosis factor receptors (TNF-R, and TNF-R2) and nerve growth factor receptors (NGF-R), CD40, OX40, CD30, CD27, and 4-1BB. Several members of this family have been shown to regulate or induce cell death (TNF-R1 and TNF-R2). A 74kDa member of this family protein named RIP (Receptor Interacting Protein) contains an N-terminal region with homology to protein kinases and a C-terminal region containing a cytoplasmic “death domain” present in both Fas and TNF-R1. Both Fas and RIP have been shown to require this death domain to induce apoptosis and overexpression of RIP has been shown to induce cell death in transfected cells.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen RIP
Applications Western Blot
Classification Monoclonal
Clone 38
Concentration 250μg/mL
Conjugate Unconjugated
Description Receptor Interacting Protein
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.