missing translation for 'onlineSavingsMsg'
Learn More

Smad2/3 Mouse, Unlabeled, Clone: 18, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610843

 View more versions of this product

Product Code. 15845669

  • ONLINE EXCLUSIVE PRICE
    336.00 €
    323.14 € / 1 set
    Save 12.86 €
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The transforming growth factor β (TGFβ)/activin/BMP family of growth factors plays a diverse and important role in growth, development, and differentiation. These growth factors act through their binding to heteromeric plasma membrane receptor protein kinases which, upon ligand binding, become activated and trigger an intracellular signaling cascade. Specifically, receptor activation induces the translocation of a set of conserved proteins named Smads (Sma- and Mad-related proteins) to the nucleus, resulting in gene activation. Smad2 is a ubiquitously expressed protein of 58kDa that is phosphorylated and translocated to the nucleus in response to TGFβ, but not BMP. The overall response to TGFβ is growth inhibition. The Smad2 gene is located in chromosome 18q21.1 which is often absent in several human cancers. Furthermore, some missense mutations on the Smad2 gene were identified in colorectal carcinomas, suggesting Smad2 may function as a tumor suppressor in normal cells.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Smad2/3
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Mouse Smad2 aa. 142-263
150 μg
Cell Biology
Canine, Human, Mouse, Rat
IgG1 κ
Western Blot
18
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.