missing translation for 'onlineSavingsMsg'
Learn More

SRP54 Mouse, Unlabeled, Clone: 30, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610940

 View more versions of this product

Product Code. 15835719

  • ONLINE EXCLUSIVE PRICE
    220.00 €
    210.79 € / 1 set
    Save 9.21 €
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Ribosomes that exist freely in the cytosol or those attached to the ER are intrinsically the same in their translational properties. ER-bound ribosomes are responsible for the production of secretory proteins and integral ER, Golgi, lysosomal, and plasma membrane spanning proteins. Such proteins contain signal sequences that direct their synthesis to the ER membrane. As the nascent polypeptide emerges from the ribosome, a signal recognition particle (SRP) binds to the signal sequence and serves to couple the ribosome to the protein-translocating machinery in the ER membrane. Although the SRP is a 325kDa ribonucleoprotein, its 54kDa subunit (SRP54) mediates interaction with, and targeting of, the nascent protein to the ER. Via its C-terminal M-domain, SRP54 associates with the nascent protein and inhibits its elongation. This complex binds to the SRP receptor on the ER, the ribosome is delivered to the translocation machinery, SRP is released, and elongation resumes. Targeting and insertion are tightly coupled to a GTPase cycle that involves SRP54 and SRP receptor. Although the mechanisms are unclear, release of SRP from the ER-bound complex requires GTP hydrolysis.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

SRP54
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human SRP54 aa. 262-476
50 μg
Cell Biology
Canine, Frog, Human, Mouse, Rat
IgG1
Western Blot
30
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.