missing translation for 'onlineSavingsMsg'
Learn More

anti-STAT1 Matched Antibody Pair, Abnova

Product Code. 16049488
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16049488 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16049488 Supplier Abnova Supplier No. H00006772AP21.1Set
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Rabbit MaxPab™ affinity purified polyclonal anti-STAT1 (100μg); Mouse purified polyclonal anti-STAT1 (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.

The protein encoded by this gene is a member of the STAT protein family. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein can be activated by various ligands including interferon-α, interferon-γ, EGF, PDGF and IL6. This protein mediates the expression of a variety of genes, which is thought to be important for cell viability in response to different cell stimuli and pathogens. Two alternatively spliced transcript variants encoding distinct isoforms have been described.

ELISA

Specifications

Antigen STAT1
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human STAT1.
Gene STAT1
Gene Alias DKFZp686B04100/ISGF-3/STAT91
Gene Symbols STAT1
Host Species Mouse, Rabbit
Purification Method Affinity Purified
Quantity 1 Set
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 6772
Includes Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-STAT1 (100 μg) 2. Detection antibody: mouse purified polyclonal anti-STAT1 (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.