missing translation for 'onlineSavingsMsg'
Learn More

Stat5 Mouse, Unlabeled, Clone: 89, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610191

 View more versions of this product

Product Code. 15865319

  • ONLINE EXCLUSIVE PRICE
    226.00 €
    216.14 € / 1 set
    Save 9.86 €
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Stat5, also known as mammary gland factor (MGF) has been shown to play a critical role in the lactogenic hormone response and has been well characterized in mammary epithelial cells. The SH3 domain of MGF shows extensive homology with the SH2 domains of Stat1 and Stat2. This homology has led to its classification as a member of the Stat family. The Stat proteins function both as cytoplasmic signal transducers and as activators of transcription. Stat (Signal transducer and activators of transcription) proteins are critical mediators of the biologic activity of cytokines, including interleukins, interferons, erythropoietin, and growth factors. Ligand-receptor interaction leads to activation of constitutively associated JAK family kinases and subsequent recruitment and activation of Stat proteins by tyrosine phosphorylation. Active Stat proteins then move to the nucleus to promote transcription of cytokine-inducible genes. Stat5a has been shown to be involved in lactogenesis and mammary development, while Stat5b has been shown to be involved in growth hormone signaling and to play a role in liver gene expression. Stat5 activity is tightly regulated throughout gestation, lactation, and post-lactation.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Stat5
Monoclonal
250μg/mL
MGF
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
89
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50 μg
Cell Biology
Canine, Human, Mouse, Rat
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.