Learn More
TEF-1 Mouse, Unlabeled, Clone: 31, BD
Mouse Monoclonal Antibody
186.18 € - 328.70 €
Specifications
| Antigen | TEF-1 |
|---|---|
| Clone | 31 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15835709
|
BD Biosciences
610922 |
50 μg |
194.00 € 186.18 € / 1 set Save 7.82 € 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15845709
|
BD Biosciences
610923 |
150 μg |
342.00 € 328.70 € / 1 set Save 13.30 € 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TEF-1 (transcription enhancer factor-1) is a member of a family of transcription factors that contain an evolutionarily conserved DNA binding domain (DBD), TEA/ATTS. TEF-1 was identified as a HeLa cell transcriptional activator that bound, via its TEA domain, to the GT-IIC and Sph enhancer elements. Comparison of various enhancers indicates that TEF-1 binds to highly degenerate DNA sequences and cooperatively interacts with tandem repeats of its binding sites, which increases its enhancer activity. However, the DNA binding activities of TEF-1 are modulated by sequences outside the DBD. The N-terminal basic portion of TEF-1 contains the DBD. The C-terminal portion contains the transactivation domain and a zinc finger-like motif that is thought to be involved in protein dimerization or interaction with additional transcription factors. TEF-1 induced transcription requires a limiting transcriptional intermediary factor (TIF) and a TATA-binding protein associated factor (hTAFIIs). Although TEF-1 is expressed ubiquitously during embryogenesis, its expression is limited to adult kidney, heart, brain, skeletal muscle, and lungs. Disruption of murine TEF-1 gene results in fetal death.
Immunofluorescence, Western Blotting
Specifications
| TEF-1 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 31 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Human TEF-1 aa. 86-199 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.