missing translation for 'onlineSavingsMsg'
Learn More

TIAR Mouse, Unlabeled, Clone: 6, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610352

Product Code. 15895409

  • ONLINE EXCLUSIVE PRICE
    187.00 €
    178.69 € / 1 set
    Save 8.31 €
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TIAR is an RNA-binding protein related to TIA-1. Both proteins are candidate effectors of apoptotic cell death and consist of three N-terminal RNA-recognition motifs (RRM) and a C-terminal protein-interaction domain (PID). Unlike TIA-1, which is localized in the granules of cytotoxic lymphocytes, TIAR is predominantly found in the nucleus of many different cell types. Two related isoforms (42 kDa and 50 kDa) of TIAR have been identified. It has been demonstrated that TIAR can trigger DNA fragmentation in permeabilized thymocytes, suggesting that it may be an effector of apoptotic cell death. During Fas-mediated apoptosis, TIAR shows a rapid translocation from the nucleus tothe cytoplasm. This redistribution of TIAR appears to be a specific event and possibly a general feature of the apoptotic program.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

TIAR
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50 μg
Cell Biology
Canine, Frog, Human, Mouse, Rat
IgG1
Western Blot
6
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.