Learn More
TNF Rat anti-Mouse, Alexa Fluor 488, Clone: MP6-XT22, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 557719
Description
The MP6-XT22 antibody reacts with mouse tumor necrosis factor (TNF, also known as TNF-α). The immunogen used to generate this hybridoma was recombinant mouse TNF. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: MP6-XT22
Isotype: IgG1
Cross Reactivity: Mouse
Flow Cytometry, Intracellular Staining
Specifications
| TNF | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant Mouse TNF | |
| 0.1 mg | |
| Immunology | |
| Mouse | |
| IgG1 |
| Flow Cytometry | |
| MP6-XT22 | |
| Alexa Fluor 488 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.