missing translation for 'onlineSavingsMsg'
Learn More

anti-TNFRSF14 Matched Antibody Pair, Abnova™

Product Code. 16182874
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16182874 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16182874 Supplier Abnova Supplier No. H00008764AP41.1Set
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Capture antibody: mouse monoclonal anti-TNFRSF14 (100μg); Detection antibody: biotinylated mouse monoclonal anti-TNFRSF14, IgG1 Kappa (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.

This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TNFRSF14.

Host Species: Mouse, Mouse
Species Reactivity: Hu

ELISA Pair (Recombinant Protein)

Specifications

Antigen TNFRSF14
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TNFRSF14.
Gene TNFRSF14
Gene Alias ATAR/HVEA/HVEM/LIGHTR/TR2
Gene Symbols TNFRSF14
Host Species Mouse
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8764
Includes Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-TNFRSF14 (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-TNFRSF14, IgG1 Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.