missing translation for 'onlineSavingsMsg'
Learn More

anti-TP53, CSNK1E Protein Protein Interaction Antibody Pair, Abnova™

Product Code. 16111144
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16111144 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16111144 Supplier Abnova Supplier No. DI0133.1Set

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

TP53 rabbit purified polyclonal antibody (20μg); CSNK1E mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols

This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the TP53 protein, and the other against the CSNK1E protein for use in in situ Proximity Ligation Assay.

Host Species: Rabbit, Mouse
Species Reactivity: Hu

In situ PLA

Specifications

Antigen TP53
Applications In situ PLA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Gene TP53
Gene Alias FLJ92943/LFS1/TRP53/p53
Gene Symbols TP53
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7157/1454
Includes Antibody pair set content: 1. TP53 rabbit purified polyclonal antibody (20 ug) 2. CSNK1E mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Type TP53 & CSNK1E Protein Protein Interaction Antibody Pair
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.