Learn More
anti-TREM1 Matched Antibody Pair, Abnova
Brand: Abnova H00054210-AP21.1Set
Includes:
- 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-TREM1 (100μg)
- 2. Detection antibody: mouse purified polyclonal anti-TREM1 (20μg)
- Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
Monocyte/macrophage- and neutrophil-mediated inflammatory responses can be stimulated through a variety of receptors, including G protein-linked 7-transmembrane receptors (e.g., FPR1; MIM 136537), Fc receptors (see MIM 146790), CD14 (MIM 158120) and Toll-like receptors (e.g., TLR4; MIM 603030), and cytokine receptors (e.g., IFNGR1; MIM 107470). Engagement of these receptors can also prime myeloid cells to respond to other stimuli. Myeloid cells express receptors belonging to the Ig superfamily, such as TREM1, or to the C-type lectin superfamily. Depending on their transmembrane and cytoplasmic sequence structure, these receptors have either activating (e.g., KIR2DS1; MIM 604952) or inhibitory functions (e.g., KIR2DL1; MIM 604936).
- Species Reactivity: Human
ELISA
Specifications
| TREM1 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TREM1. | |
| TREM-1 | |
| Mouse, Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-TREM1 (100 μg) 2. Detection antibody: mouse purified polyclonal anti-TREM1 (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Unconjugated | |
| TREM1 | |
| TREM1 | |
| NA; TREM1 (NP_061113.1, 1 a.a. ∽ 234 a.a) full-length human protein. | |
| 1 Set | |
| Signal Transduction | |
| 54210 | |
| Human |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.