Learn More
WNT16 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534318
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Mouse, Rat, Hamster (100%) Gorilla, Monkey, Marmoset (94%) Gibbon, Elephant, Panda, Dog, Horse, Turkey (88%).
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It contains two transcript variants diverging at the 5' termini. These two variants are proposed to be the products of separate promoters and not to be splice variants from a single promoter. They are differentially expressed in normal tissues, one of which (variant 2) is expressed at significant levels only in the pancreas, whereas another one (variant 1) is expressed more ubiquitously with highest levels in adult kidney, placenta, brain, heart, and spleen. WNT15 is also a ligand for members of the frizzled family of seven transmembrane receptors.Specifications
WNT16 | |
Polyclonal | |
Unconjugated | |
Wnt16 | |
E130309I19Rik; Protein Wnt-16; wingless-related MMTV integration site 16; wingless-type MMTV integration site family member 16; wingless-type MMTV integration site family member 16b; wingless-type MMTV integration site family, member 16; Wnt family member 16; Wnt16 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
500047, 51384, 93735 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9QYS1, Q9UBV4 | |
Wnt16 | |
Synthetic 16 amino acid peptide from internal region of human WNT16. | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |