missing translation for 'onlineSavingsMsg'
Learn More

Antibiotic-5d, MedChemExpress

Product Code. 16474466
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16474466 100 mg 100mg
16465917 5 mg 5mg
16459736 50 mg 50mg
16443188 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16474466 Supplier Medchem Express Supplier No. HY100833
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Antibiotic-5d is a synthesis and antimicrobial compound.

Antibiotic-5d is a synthesis and antimicrobial compound.

Specifications

Chemical Name or Material Antibiotic-5d
Physical Form Solid
CAS 251349-54-9
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C13H18N2O4S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 50 mg/mL (167.58 mM; Need ultrasonic)
SMILES O=C(C1=CSC([C@H]2N(C(OC(C)(C)C)=O)CCC2)=N1)O
Molecular Weight (g/mol) 298.36
Formula Weight 298.36
Percent Purity 98.35%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.