Learn More
Invitrogen™ Telazorlimab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559806
 View more versions of this product
Product Code. 30282373
  • 1204.00 € / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Telazorlimab is a monoclonal antibody that targets and inhibits the activity of the cytokine interleukin-4 (IL-4) and interleukin-13 (IL-13), thereby modulating the immune response.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Telazorlimab Humanized
Recombinant Monoclonal
Unconjugated
GBR 830
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized