missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AOC3 Recombinant Human Monoclonal Antibody (3D3)

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560763
This item is not returnable.
View return policy
Description
AOC3 Recombinant Monoclonal Antibody for IHC (P), ELISA
Copper amine oxidases catalyze the oxidative conversion of amines to aldehydes in the presence of copper and quinone cofactor. The product is a major protein on the adipocyte plasma membrane. It has adhesive properties and also has functional monoamine oxidase activity. A pseudogene for this gene has been discribed and is located approximately 9-kb downstream.
Specifications
| AOC3 | |
| Recombinant Monoclonal | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q16853 | |
| AOC3 | |
| Recombinant Human AOC3 protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG4 |
| ELISA, Immunohistochemistry (Paraffin) | |
| 3D3 | |
| Unconjugated | |
| AOC3 | |
| amine oxidase, copper containing 3; amine oxidase, copper containing 3 (vascular adhesion protein 1); Aoc3; Copper amine oxidase; HPAO; Membrane primary amine oxidase; placenta copper monamine oxidase; semicarbazide-sensitive amine oxidase; SSAO; VAP1; VAP-1; Vascular adhesion protein 1; VP97 | |
| Human | |
| Affinity chromatography | |
| RUO | |
| 8639 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction