missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP1M2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-55000
This item is not returnable.
View return policy
Description
AP1M2 Polyclonal specifically detects AP1M2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| AP1M2 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| Adaptor protein complex AP-1 mu-2 subunit, Adaptor-related protein complex 1 mu-2 subunit, adaptor-related protein complex 1, mu 2 subunit, AP-1 complex subunit mu-2, AP1-mu2, AP-mu chain family member mu1B, Clathrin assembly protein complex 1 medium chain 2, clathrin coat assembly protein AP47 2, clathrin coat associated protein AP47 2, clathrin-associated adaptor medium chain mu2, golgi adaptor AP-1 47 kDa protein, Golgi adaptor HA1/AP1 adaptin mu-2 subunit, HA1 47 kDa subunit 2, HSMU1B, MU1B, MU-1B, mu1B-adaptin, mu2, Mu-adaptin 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 10053 | |
| Human | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| AP1M2 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MPLLVQREEEGALAPLLSHGQVHFLWI | |
| 100 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction