Learn More
Invitrogen™ AP2 alpha Recombinant Rabbit Monoclonal Antibody (24GB2485)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557769
 View more versions of this product
Product Code. 30275089
  • 198.00 € / 20µL
Description
AP2 alpha Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
TFAP2A is a sequence-specific DNA-binding protein that interacts with inducible viral and cellular enhancer elements to regulate transcription of selected genes. AP-2 factors bind to the consensus sequence 5'-GCCNNNGGC-3' and activate genes involved in a large spectrum of important biological functions including proper eye, face, body wall, limbs and neural tube development. They also suppress a number of genes including MCAM/MUC18, C/EBP alpha and MYC. TFAP2A is the only AP-2 protein required for early morphogenesis of the lens vesicle.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AP2 alpha
Recombinant Monoclonal
Unconjugated
TFAP2A
activating enhancer-binding protein 2 alpha; activating enhancer-binding protein 2-alpha; Activator protein 2; activator protein 2 alpha variant 4; activator protein 2 alpha variant 6; activator protein 2 alpha variant 7; AP2; AP-2; Ap-2 (a); AP-2 transcription factor; AP2alpha; AP-2alpha; AP2-alpha; Ap2tf; BOFS; ERF-1; FLJ51761; OTTHUMP00000214240; OTTHUMP00000214243; Tcfap2a; TFAP2; TFAP2A; transcription factor AP2; transcription factor AP-2 alpha; transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha); transcription factor AP-2, alpha; transcription factor AP-2-alpha; transcription factor AP-2-alpha-like; transcription factor; AP-2
Rabbit
Affinity Chromatography
RUO
306862, 7020
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2485
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P05549, P58197
TFAP2A
A synthesized peptide derived from human AP2 alpha (aa 100-140).
20 μL
Primary
Human, Rat