missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APCDD1L Antibody (Z35P1F10*C4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
190.00 € - 351.00 €
Specifications
| Antigen | APCDD1L |
|---|---|
| Clone | Z35P1F10*C4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18627927
|
Novus Biologicals
NBP2-50107-0.025ml |
0.025 mL |
190.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18657237
|
Novus Biologicals
NBP2-50107 |
0.1 mL |
351.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
APCDD1L Monoclonal antibody specifically detects APCDD1L in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| APCDD1L | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cellular Signaling | |
| PBS | |
| 164284 | |
| IgG | |
| Protein A purified |
| Z35P1F10*C4 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Adenomatosis polyposis coli down-regulated 1 protein-like, adenomatosis polyposis coli down-regulated 1-like, FLJ90166, MGC126807, MGC126809, protein APCDD1-like, RP4-685L9.2 | |
| Peptide Sequence -KRPTSYQA (immunogen peptide is identical in sheep, bovine, porcine, equine and chicken) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title