missing translation for 'onlineSavingsMsg'
Learn More

Apigenin, MedChemExpress

Product Code. 16484907
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484907 50 mg 50mg
16499146 100 mg 100mg
16469208 10 mg 10mg
16431516 200 mg 200mg
16422396 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16484907 Supplier Medchem Express Supplier No. HYN1201
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Apigenin (4',5,7-Trihydroxyflavone) is a competitive CYP2C9 inhibitor with a Ki of 2 μM.

Apigenin (4',5,7-Trihydroxyflavone) is a competitive CYP2C9 inhibitor with a Ki of 2 μM.

Specifications

Chemical Name or Material Apigenin
CAS 520-36-5
Color Green Yellow
Purity Grade Notes Research
Molecular Formula C15H10O5
Quantity 50 mg
Synonym 4',5,7-Trihydroxyflavone Apigenol C.I. Natural Yellow 1
Solubility Information DMSO : 83.33 mg/mL (308.36 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OC1=C2C(OC(C3=CC=C(O)C=C3)=CC2=O)=CC(O)=C1
Molecular Weight (g/mol) 270.24
Formula Weight 270.24
Percent Purity 96.92%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.