missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apo-Transferrin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-43440
This item is not returnable.
View return policy
Description
Apo-Transferrin Polyclonal antibody specifically detects Apo-Transferrin in Human samples. It is validated for Western Blot
Specifications
| Apo-Transferrin | |
| Polyclonal | |
| Unconjugated | |
| 0.01 M PBS, pH 7.4, 50% glycerol | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 7037 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 0.01-2ug/mL | |
| CD71, p90, T9, TFR, TFR1, TFRC, TR, transferrin receptor (p90, CD71), TRFR | |
| Full-length Apo-Transferrin | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction