Learn More
ApoA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA589682
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: DEPPQSPWDR VKDLATVYVD VLKDSGRDYV SQFEGSALGK QLNLKLLDNW DSVTSTFSKL REQLGPVTQE FWDNLEKETE GLRQEMSKDL EEVKAKVQPY LDDFQKKWQE EMELYRQKVE PLRAELQEGA RQKLHELQEK LSPLGEEMRD RARAHVDALR THLAPYSDEL RQRLAARLEA LKENGGARLA EYHAKATEHL STLSEKAKPA LEDLRQGLLP VLESFKVSFL SALEEYTKKL NTQ; Positive Samples: Mouse lung, mouse liver, mouse small intestine, rat lung, rat liver
Apolipoprotein A-1 (apo A-1) in human plasma is a major protein component of high density lipoprotein (HDL). The molecular mass is 24 kDa. Apo A-1 is a cofactor for enzymes involved in the normal turnover of cholesterol and is produced in the liver. Plasma concentration is 270-380 mg/100 mL.Specifications
ApoA1 | |
Polyclonal | |
Unconjugated | |
APOA1 | |
1-Sep, 2-Sep, Alp-1, Apo a, APO A1, apo A-1, APO I, apo(a), APOA1, Apoa-1, ApoAI, Apo-AI, apoA-I, Apolipoprotein, Apolipoprotein A1, apolipoprotein A-1, Apolipoprotein AI, apolipoprotein A-I, apolipoprotein A-I precursor, Apolipoprotein A-I(1-242), Brp-14, Ltw-1, Lvtw-1, MGC102525, MGC117399, MGC124929, preproapolipoprotein A-I, prepro-apolipoprotein-AI, ProapoA-I, Proapolipoprotein A-I, Sep-1, Sep2, Sep-2, Truncated apolipoprotein A-I, Udpgt, Ugt1, Ugt1a7 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
11806, 25081 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide, pH 7.3 | |
P04639, Q00623 | |
APOA1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 25-267 of human APOA1 (NP_0000301) | |
100 μL | |
Primary | |
Mouse, Rat, Rat | |
Antibody | |
IgG |