missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOBEC3B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99174-50ul
This item is not returnable.
View return policy
Description
APOBEC3B Polyclonal antibody specifically detects APOBEC3B in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| APOBEC3B | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-4 uL/mg of lysate | |
| APOBEC1L, apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3B, ARCD3, ARP4, bK150C2.2, cytidine deaminase, DJ742C19.2, EC 3.5.4, EC 3.5.4.-, FLJ21201, phorbolin 3, Phorbolin-1-related protein, phorbolin-2/3, PHRBNLphorbolin 2, probable DNA dC->dU-editing enzyme APOBEC-3B | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human APOBEC3B. | |
| 50 μg | |
| Immunology | |
| 9582 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction