missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein A II Mouse anti-Human, Clone: 1G1A2, Proteintech
Mouse Monoclonal Antibody
140.00 € - 399.00 €
Specifications
Antigen | Apolipoprotein A II |
---|---|
Clone | 1G1A2 |
Concentration | 0.8 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16888955
|
Proteintech
66773-1-IG-20UL |
20 μL |
140.00 €
20µL |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16878955
|
Proteintech
66773-1-IG-150UL |
150 μL |
399.00 €
150µL |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
At least 9 distinct polymorphic forms of apolipoproteins are known. The apolipoproteins act as stabilizers of the intact lipoprotein particles. Quantitative measurements of HDL, LDL, and VLDL particles of human serum are often used to estimate an individual′s relative risk of coronary heart disease. In addition, quantitative immunological measurements of certain apolipoproteins ( especially A-1 and B) have been suggested to be more accurate estimators of coronary heart disease that measurements of lipoprotein particles (especially HDL and LDL). The apolipoproteins are derived from human plasma by density gradient ultracentrifugation and HPLC.Spezifikation
Apolipoprotein A II | |
0.8 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Apo AII, ApoA II, APOA2, apolipoprotein A II, Apolipoprotein A2 | |
APOA2 | |
IgG2b | |
Protein A | |
Antibody |
1G1A2 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P02652 | |
336 | |
Apolipoprotein A II Fusion Protein Ag9863 | |
Primary | |
-20°C | |
APOA2 |