missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein A-IV/ApoA4 Antibody (CL0467) [Alexa Fluor™ 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52931AF405
This item is not returnable.
View return policy
Description
Apolipoprotein A-IV/ApoA4 Monoclonal antibody specifically detects Apolipoprotein A-IV/ApoA4 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Apolipoprotein A-IV/ApoA4 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| apo-AIV, ApoA-IV, Apolipoprotein A4, apolipoprotein A-IV, MGC142154, MGC142156 | |
| This antibody was developed using a recombinant protein derived from P06727, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 337 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL0467 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction