missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein E/ApoE Antibody (WUE-4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 22 publications
Brand: Novus Biologicals NB110-60531
This item is not returnable.
View return policy
Description
Apolipoprotein E/ApoE Monoclonal specifically detects Apolipoprotein E/ApoE in Human, Mouse, Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Intracellular), CyTOF-ready.
Specifications
| Apolipoprotein E/ApoE | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 2 ug/ml, Flow Cytometry 1 ug per million cells, ELISA 1:100-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin, CyTOF-ready | |
| Apo-E, apolipoprotein E, apolipoprotein E3, late onset), MGC1571 | |
| Mouse | |
| 36 kDa | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Cognition and Behavior, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 348 | |
| Human, Mouse, Rat (Negative) | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| WUE-4 | |
| Unconjugated | |
| P02649 | |
| APOE | |
| Purified human ApoE [UniProt# P02649] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human and Mouse. It does not appear to react with rat brain tissue. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction