missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein E/ApoE Antibody (WUE-4), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60531PE
This item is not returnable.
View return policy
Description
Apolipoprotein E/ApoE Monoclonal specifically detects Apolipoprotein E/ApoE in Human, Mouse, Rat (Negative) samples. It is validated for Flow Cytometry, Flow (Intracellular).
Specifications
| Apolipoprotein E/ApoE | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| APOE | |
| Purified human ApoE [UniProt# P02649] | |
| 0.1 mL | |
| Primary | |
| This antibody detects ApoE2, ApoE3 and ApoE4 (PMID 2466929). | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| WUE-4 | |
| Flow Cytometry, Flow (Intracellular) | |
| Apo-E, apolipoprotein E, apolipoprotein E3, late onset), MGC1571 | |
| Mouse | |
| Protein G purified | |
| Alzheimers Research, Apoptosis, Cancer, Cholesterol Metabolism, Cognition and Behavior, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 348 | |
| Human, Mouse, Rat (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction