Learn More
Apolipoprotein L1 Antibody (CL0173) [Alexa Fluor™ 647], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52871AF647
Product Code. 30293382
  • 737.00 € / 0.10mL
Description
Apolipoprotein L1 Monoclonal antibody specifically detects Apolipoprotein L1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Apolipoprotein L1
Monoclonal
Alexa Fluor 647
ApoL, APO-L, ApoL-I, Apolipoprotein L, apolipoprotein L, 1, apolipoprotein L1, Apolipoprotein L-I, FSGS4
This antibody was developed using a recombinant protein derived from O14791, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cardiovascular Biology, Endocrinology
8542
Store at 4⪚C in the dark.
IgG1
Western Blot
CL0173
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified