missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Apolipoprotein O Monoclonal Antibody (2F1)
GREENER_CHOICE

Product Code. 11385463
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11385463 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11385463 Supplier Invitrogen™ Supplier No. MA515493

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA5-15493 targets ApoO in indirect ELISA, IHC and WB applications and shows reactivity with Human and mouse samples. The MA5-15493 immunogen is purified recombinant fragment of ApoO expressed in E. Coli. . MA5-15493 detects ApoO which has a predicted molecular weight of approximately 22.2kDa.

This gene is a member of the apolipoprotein family. Members of this protein family are involved in the transport and metabolism of lipids. The encoded protein associates with HDL, LDL and VLDL lipoproteins and is characterized by chondroitin-sulfate glycosylation. This protein may be involved in preventing lipid accumulation in the myocardium in obese and diabetic patients. Alternative splicing results in multiple transcript variants. Pseudogenes of this gene are found on chromosomes 3, 4, 5, 12 and 16.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein O
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2F1
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene APOO
Gene Accession No. Q9BUR5, Q9DCZ4
Gene Alias 0610008C08Rik; 1110019O03Rik; Apolipoprotein O; APOO; brain my025; FAM121B; family with sequence similarity 121B; MGC4825; Mic23; MIC26; MICOS complex subunit Mic23; MICOS complex subunit MIC26; My025; MYO25; Protein FAM121B; RGD1565289; UNQ1866/PRO4302
Gene Symbols APOO
Host Species Mouse
Immunogen Purified recombinant fragment of ApoO expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68316, 79135
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.