missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apoptosis enhancing nuclease Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-87016
This item is not returnable.
View return policy
Description
Apoptosis enhancing nuclease Polyclonal specifically detects Apoptosis enhancing nuclease in Human samples. It is validated for Western Blot.
Specifications
| Apoptosis enhancing nuclease | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 64782 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| apoptosis enhancing nuclease, EC 3.1, EC 3.1.13.1, FLJ12484, FLJ12562, interferon stimulated exonuclease gene 20kDa-like 1, Interferon-stimulated 20 kDa exonuclease-like 1, ISG20L1apoptosis-enhancing nuclease, pp12744 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of human Apoptosis enhancing nuclease | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction