missing translation for 'onlineSavingsMsg'
Learn More

Aposcopolamine, TRC

Product Code. 30137903
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137903 25 mg 25mg
17959939 5 mg 5mg
18070520 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137903 Supplier Toronto Research Chemicals Supplier No. TRCA72900025MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 535-26-2
Molecular Formula C17 H19 N O3
Molecular Weight (g/mol) 285.34
Synonym Benzeneacetic acid, α-methylene-, (1α,2β,4β,5α,7β)-9-methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl ester, 1αH,5αH-Tropan-3α-ol, 6β,7β-epoxy-, atropate (ester) (8CI), Aposcopolamine (6CI,7CI), Benzeneacetic acid, α-methylene-, 9-methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl ester, (1α,2β,4β,5α,7β)-, 3-Oxa-9-azatricyclo[3.3.1.02,4]nonane, benzeneacetic acid deriv., Apohyoscine, Aposcopolamin, (1R,2R,4S,5S,7s)-9-Methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl 2-phenylprop-2-enoate, Hyoscine Hydrobromide Imp. C (EP)
IUPAC Name (1R,2R,4S,5S,7s)-9-Methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl 2-Phenylprop-2-enoate
SMILES CN1[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)C(=C)c4ccccc4

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C17H19NO3/c1-10(11-6-4-3-5-7-11)17(19)20-12-8-13-15-16(21-15)14(9-12)18(13)2/h3-7,12-16H,1,8-9H2,2H3/t12-,13-,14+,15-,16+
Formula Weight 285.1365
Percent Purity >95%
Chemical Name or Material (1R,2R,4S,5S,7s)-9-Methyl-3-oxa-9-azatricyclo(3.3.1.02,4)non-7-yl 2-Phenylprop-2-enoate (Apohyoscine)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.