Learn More
APPL1 Mouse, anti-Human, CoraLite Plus 488, Clone: 1B7B11, Proteintech
Mouse Monoclonal Antibody
145.00 € - 450.00 €
Specifications
| Antigen | APPL1 |
|---|---|
| Clone | 1B7B11 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18037415
|
Proteintech
CL488-68195-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18029495
|
Proteintech
CL488-68195-100UL |
100 μL |
450.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The APPL1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human APPL1. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The APPL1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| APPL1 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 26060 | |
| IgG2b | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1B7B11 | |
| Immunofluorescence | |
| Liquid | |
| Human, Mouse, Rat, Rabbit, Pig | |
| APPL, APPL1, Dip13 alpha, DIP13A, DIP13alpha, KIAA1428 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.