missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 96-Well Clear GPLE Reaction Plates with Barcode

Product Code. 15352476 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
20 Plates
500 Plates
Unit Size:
Pack of 20
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15352476 20 Plates Pack of 20
15362476 500 Plates Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15352476 Supplier Applied Biosystems™ Supplier No. 4483348
Simplify Your Gene Expression Workflow
Use promo code 27954
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Twenty 96-Well plates

Each lot is certified DNA, RNAse, and PCR inhibitor-free

  • For Laboratory Use
  • Optimized for performance with Applied Biosystems instruments
  • High mechanical stability and flatness for complete workflow needs
  • Polypropylene and high temperature molding process helps prevent DNA, RNA, and enzymes from binding, while polycarbonate offers mechanical stability, flatness, and high durability
  • ANSI™/SBS-compliant construction
  • Ideal choice for use with automation/robotics and multi-instrument experiments (like thermal cycler, real-time thermal cycler, genetic analyzer)
  • Thin-wall construction allows for snug fit to thermal blocks with high heat transfer from thermal block to sample
  • Easy-to-read, black, well-identification text helps to minimize pipetting mistakes and improves sample preparation productivity

  • Platform: 2720 Thermal Cycler, 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, 7300 System, 7500 System, 7900HT System, GeneAmp™ 9700, ProFlex™ PCR System, QuantStudio™, QuantStudio Dx, SimpliAmp™ Thermal Cycler, Veriti™ Dx Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA 7 System

Fast PCR, PCR, PCR & Real-Time PCR, Routine PCR

TRUSTED_SUSTAINABILITY

Specifications

Barcode Includes Barcode
Certifications/Compliance Certified DNA/RNase Free
Content And Storage 20 96-Well plates. Store at room temperature.
Packaging 20 Plates
Product Line MicroAmp
Color Clear, Optical
Product Type Microplate
For Use With (Equipment) Thermal Cycler, Genetic Analyzer, DNA Analyzer
Skirt Style Semi
Material Polycarbonate, Polypropylene
Volume (Metric) Well 0.3 mL
No. of Wells 96
Volume (Metric) Working 0.2 mL
Quantity 20 Plates
Reaction Speed Fast, Standard
Show More Show Less

For Laboratory Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.