missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ Optical Film Compression Pad

Product Code. 10065764 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
5 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10065764 5 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10065764 Supplier Applied Biosystems™ Supplier No. 4312639
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

5 Optical Cover Compression Pads

For use with adhesive films

Designed to support a proper thermal seal between the thermal cycler and reaction plates when using adhesive films.

See user's manual or package insert for limited label license, and trademark information.

PCR and Real-Time PCR, Real Time PCR (qPCR)

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Optical
Quantity 5 Ea.
For Use With (Application) PCR
Content And Storage 5 Optical Cover Compression Pads
For Use With (Equipment) 2720 Thermal Cycler, 3100 Genetic Analyzer, 3700 Genetic Analyzer, 6100 Thermal Cycler, 7000 System, 7300 System, 7500 System, 7900HT System, GeneAmp 9700, Veriti Thermal Cycler, 2720 Thermal Cycler
Shipping Condition Room Temperature
Product Type Compression Pad
Product Line MicroAmp
Plate Compatibility 96-well Plate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.