Learn More
Invitrogen™ Aprataxin Recombinant Rabbit Monoclonal Antibody (24GB6830)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556966
 View more versions of this product
Product Code. 30274837
  • 146.00 € / 20µL
Description
Aprataxin Recombinant Monoclonal Antibody for Western Blot
This gene encodes a member of the histidine triad (HIT) superfamily, some of which have nucleotide-binding and diadenosine polyphosphate hydrolase activities. The encoded protein may play a role in single-stranded DNA repair. Mutations in this gene have been associated with ataxia-ocular apraxia. Multiple transcript variants encoding distinct isoforms have been identified for this gene, however, the full length nature of some variants has not been determined.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Aprataxin
Recombinant Monoclonal
Unconjugated
APTX
2410016G21Rik; AA388047; AOA; AOA1; Aprataxin; aprataxin isoform a; Aptx; APTX protein; AXA1; EAOH; EOAHA; FHA-HIT; Forkhead-associated domain histidine triad-like protein; forkhead-associated domain histidine-triad like protein
Rabbit
Affinity chromatography
RUO
54840
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB6830
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q7Z2E3
APTX
A synthesized peptide derived from human ApRataxin (aa 147-160).
20 μL