missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AQPEP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562765
This item is not returnable.
View return policy
Description
Immunogen sequence: IPYPIKDVVL CYGIALGSDK EWDILLNTYT NTTNKEEKIQ LAYAMSCSKD PWILNRYMEY AISTSPFTSN ETNIIEVVAS SEVGRYVA Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 61%.
Metalloprotease which may be important for placentation by regulating biological activity of key peptides at the embryo-maternal interface. On synthetic substrates it shows a marked preference for Leu-4-methylcoumaryl-7-amide (Leu-MCA) over Met-MCA, Arg-LCA and Lys-LCA. Cleaves the N-terminal amino acid of several peptides such as angiotensin-3, kisspeptin-10 and endokinin C. [UniProt].
Specifications
| AQPEP | |
| Polyclonal | |
| Unconjugated | |
| LVRN | |
| Aminopeptidase Q; APQ; AP-Q; AQPEP; CHL2 antigen; laeverin; LVRN; TAQPEP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 206338 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6Q4G3 | |
| LVRN | |
| Recombinant protein corresponding to Human AQPEP. Recombinant protein control fragment (Product #RP-100461). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction