missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin-9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12260
This item is not returnable.
View return policy
Description
Aquaporin-9 Polyclonal antibody specifically detects Aquaporin-9 in Mouse, Rat, Bovine, Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| Aquaporin-9 | |
| Polyclonal | |
| Western Blot 1:500, ELISA 1:10000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin | |
| AQP-9, Aquaglyceroporin-9, aquaporin 9, aquaporin-9, HsT17287, Small solute channel 1, SSC1aquaglyceroporin-9 | |
| Synthetic peptide taken within amino acid region 250-294 on rat Aquaporin 9 protein. | |
| 100 μg | |
| Cancer, Endocrinology, Signal Transduction | |
| 366 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo- protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide | |
| Rabbit | |
| Affinity-purified | |
| RUO | |
| Primary | |
| Mouse, Rat, Bovine, Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction