missing translation for 'onlineSavingsMsg'
Learn More

Aramite 2000 μg/mL in n-Hexane, Dr. Ehrenstorfer

Product Code. 30328664
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30328664 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328664 Supplier Dr. Ehrenstorfer Supplier No. DREGA09010391HE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aramite
CAS 140-57-8
Analyte or Component Names Aramite
Shipping Condition Room Temperature
Molecular Formula C15 H23 Cl O4 S
Concentration 2000 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C15H23ClO4S/c1-12(20-21(17)19-10-9-16)11-18-14-7-5-13(6-8-14)15(2,3)4/h5-8,12H,9-11H2,1-4H3
SMILES CC(COc1ccc(cc1)C(C)(C)C)OS(=O)OCCCl
IUPAC Name 1-(4-tert-butylphenoxy)propan-2-yl 2-chloroethyl sulfite
Solvent or Matrix n-Hexane
Molecular Weight (g/mol) 334.86
Quantity 1 mL
Formula Weight 334.1006 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.