missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARF1 Recombinant Rabbit Monoclonal Antibody (23GB3155), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553373
This item is not returnable.
View return policy
Description
ADP-ribosylation factor 1 (ARF1) is a member of the human ARF gene family. The family members encode small guanine nucleotide-binding proteins that stimulate the ADP-ribosyltransferase activity of cholera toxin and play a role in vesicular trafficking as activators of phospholipase D. The gene products, including 6 ARF proteins and 11 ARF-like proteins, constitute a family of the RAS superfamily. The ARF proteins are categorized as class I (ARF1, ARF2 and ARF3), class II (ARF4 and ARF5) and class III (ARF6), and members of each class share a common gene organization. The ARF1 protein is localized to the Golgi apparatus and has a central role in intra-Golgi transport. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| ARF1 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P84077, P84078, P84079 | |
| ARF1 | |
| A synthetic peptide of human ARF1 (70-108AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3155 | |
| Unconjugated | |
| ARF1 | |
| ADP ribosylation factor 1; ADP ribosylation factor 1 L homeolog; ADP-ribosylation factor 1; Arf1; arf1.L; arf2; wu:fb33e02; wu:fb78h01; XELAEV_18043369mg | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11840, 375, 64310 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction