missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARF3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
227.00 € - 326.00 €
Specifications
| Antigen | ARF3 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18689645
|
Novus Biologicals
NBP2-99493-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18637765
|
Novus Biologicals
NBP2-99493-50ul |
50 μg |
227.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARF3 Polyclonal antibody specifically detects ARF3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| ARF3 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.0) | |
| 377 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ADP-ribosylation factor 3, small GTP binding protein | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ARF3. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title