missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARFGAP1 Monoclonal Antibody (OTI 2C10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502635
This item is not returnable.
View return policy
Description
ArfGAP1 is a 406 amino acid containing regulatory protein with an Arf-GAP domain known to be involved in the activation of the GTPase activity of ADP ribosylation factor (ARF). ArfGAP1, a GTPase-activating protein (GAP) for the ADP ribosylation factor 1 (ARF1), promotes the hydrolysis of ARF1-bound GTP and is required for the dissociation of coat proteins from Golgi-derived membranes and vesicles thus playing an important role in membrane trafficking and/or vesicle transport. Dissociation of the coat proteins is required for the fusion of these vesicles with target compartments. The activity of this protein is stimulated by phosphoinosides and inhibited by phosphatidylcholine. Two transcript variants encoding different isoforms have been reported. Ubiquitous expression of ArfGAP1 is seen in most tissues.Specifications
| ARFGAP1 | |
| Monoclonal | |
| 0.4 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8N6T3 | |
| ARFGAP1 | |
| Full length human recombit protein of human ARFGAP1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI 2C10 | |
| Unconjugated | |
| ARFGAP1 | |
| ARF1GAP, HRIHFB2281 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55738 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction