missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arginase 1/ARG1/liver Arginase Rabbit anti-Human, Clone: 34, Novus Biologicals™
Rabbit Monoclonal Antibody
367.00 € - 552.00 €
Specifications
| Antigen | Arginase 1/ARG1/liver Arginase |
|---|---|
| Clone | 34 |
| Dilution | Western Blot 1:500-1:1000, Flow Cytometry 1:100-1:500, ELISA 1:25000-1:50000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunohistochemistry-Paraffin 1:500-1:2500 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18274847
|
Novus Biologicals
NBP2-89915-50UL |
50 μg |
367.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18244068
|
Novus Biologicals
NBP2-89915-100UL |
100 μg |
552.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Arginase 1/ARG1/liver Arginase Monoclonal specifically detects Arginase 1/ARG1/liver Arginase in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Arginase 1/ARG1/liver Arginase | |
| Western Blot 1:500-1:1000, Flow Cytometry 1:100-1:500, ELISA 1:25000-1:50000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunohistochemistry-Paraffin 1:500-1:2500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, Liver-type arginase, Type I arginase | |
| Recombinant Human Arginase 1/ARG1/liver Arginase protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 34 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cellular Markers, Chromatin Research, Lipid and Metabolism, Myeloid derived Suppressor Cell | |
| 0.2 um filtered solution in PBS | |
| 383 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title