missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arginine decarboxylase Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92284-0.1ml
This item is not returnable.
View return policy
Description
Arginine decarboxylase Polyclonal antibody specifically detects Arginine decarboxylase in Mouse, Rat samples. It is validated for Western Blot
Specifications
| Arginine decarboxylase | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| antizyme inhibitor 2, ARGDC, arginine decarboxylase, AZI2, KIAA1945, ODC antizyme inhibitor-2, ODC1L, ODC-p, ODC-paralogue, ornithine decarboxylase-like protein, ornithine decarboxylase-paralog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-360 of human AZIN2 (NP_443724.1). MAGYLSESDFVMVEEGFSTRDLLKELTLGASQATTDEVAAFFVADLGAIVRKHFCFLKCLPRVRPFYAVKCNSSPGVLKVLAQLGLGFSCANKAEMELVQHIGIPASKIICANPCKQIAQIKYAAKHGIQLLSFDNEMELAKVVKSHPSAKMVLCIATDDSHSLSCLSLKFGVSLKSCRHLLENAKKHHVEVVGVSFHIGSGCPDPQAYAQSIADARLVFEMGTELGHKMHVLDLGGGFPGTEGAKVRFEEIASVINSALDLYFPEGCGVDIFAELGRYYVTSAFTVAVSIIAKKEVLLDQPGREEENGSTSKTIVYHLDEGVYGIFNSVLFDNICPTPILQKKPSTEQPLYSSSLWGPA | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
| 113451 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction