missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ArgRS Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92835-0.1ml
This item is not returnable.
View return policy
Description
ArgRS Polyclonal antibody specifically detects ArgRS in Human, Mouse samples. It is validated for Western Blot
Specifications
| ArgRS | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| arginine-tRNA ligase, arginyl-tRNA synthetase 2, mitochondrial, Arginyl-tRNA synthetase-like, ArgRS, DALRD2, dJ382I10.6, EC 6.1.1.19, MGC14993, MGC23778, PCH6, PRO1992, probable arginyl-tRNA synthetase, mitochondrial, RARSLArginine--tRNA ligase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 17-340 of human RARS2 (NP_064716.2). LNLPPENLITSISAVPISQKEEVADFQLSVDSLLEKDNDHSRPDIQVQAKRLAEKLRCDTVVSEISTGQRTVNFKINRELLTKTVLQQVIEDGSKYGLKSELFSGLPQKKIVVEFSSPNVAKKFHVGHLRSTIIGNFIANLKEALGHQVIRINYLGDWGMQFGLLGTGFQLFGYEEKLQSNPLQHLFEVYVQVNKEAADDKSVAKAAQEFFQRLELGDVQALSLWQKFRDLSIEEYIRVYKRLGVYFDEYSGESFYREKSQEVLKLLESKGLLLKTIKGTAVVDLSGNGDPSSICTVMRSDGTSLYATRDLAAAIDRMDKYNFD | |
| 0.1 mL | |
| Endocrinology | |
| 57038 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction