missing translation for 'onlineSavingsMsg'
Learn More

ARHGAP11B Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-06142PE

Product Code. 18629867

  • 407.00 € / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ARHGAP11B Polyclonal antibody specifically detects ARHGAP11B in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ARHGAP11B
Polyclonal
Immunohistochemistry-Paraffin
B'-T, FAM7B1, Family With Sequence Similarity 7, Member B1, GAP (1-8), Rho GTPase Activating Protein 11B, Rho GTPase-Activating Protein 11B, Rho-Type GTPase-Activating Protein 11B
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human ARHGAP11B.
0.1 mL
GPCR, Signal Transduction
89839
Store at 4C in the dark.
IgG
Immunohistochemistry (Paraffin)
PE
PBS
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.