missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP17 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
292.00 € - 624.00 €
Specifications
| Antigen | ARHGAP17 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18448220
|
Novus Biologicals
NBP1-83870-25ul |
25 μL |
292.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18294486
|
Novus Biologicals
NBP1-83870 |
0.1 mL |
624.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARHGAP17 Polyclonal specifically detects ARHGAP17 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ARHGAP17 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| DKFZp564A1363, FLJ10308, FLJ13219, FLJ37567, FLJ43368, MGC87805, MST110, MSTP038, MSTP066, MSTP110, NADRIN, neuron-associated developmentally regulated protein, PP367, PP4534, Rho GTPase activating protein 17, rho GTPase-activating protein 17, RhoGAP interacting with CIP4 homologs 1, RhoGAP interacting with CIP4 homologs protein 1, Rho-type GTPase-activating protein 17, RICH-1, RICH1B, RICH1MST066, WBP15 | |
| ARHGAP17 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse | |
| Q68EM7 | |
| 55114 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KQLARLVLDWDSVRARWNQAHKSSGTNFQGLPSKIDTLKEEMDEAGNKVEQCKDQLAADMYNFMAKEGEYGKFFVTLLEA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title